Assay Details
Target Gene Details
Entrez Gene ID: | 219151 |
Gene Name: | scavenger receptor class A, member 3 |
Gene Aliases: |
APC7, C130058N24Rik, CSR, CSR1, MSLR1, MSRL1 |
Location: |
Chr.14:65919395-65953744 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Scara3 | NM_172604.3 | 6 | 2513 | NP_766192.1 |
| AK035740.1 | 1 | 653 | ||
| AK048423.1 | 6 | 2514 | BAC33332.1 | |
| BC026446.1 | 2 | 1303 | AAH26446.1 | |
| BC051636.1 | 6 | 2286 | AAH51636.2 |
More Information
Set Membership: |
|

Genomic Map