Assay Details
Target Gene Details
Entrez Gene ID: | 100038602 |
Gene Name: | predicted gene 10865 |
Gene Aliases: |
ENSMUSG00000075558 |
Location: |
Chr.15:78994955-79011041 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm10865 | NR_045746.1 | |||
| AK139534.1 | BAE24054.1 |
Target Gene Details
Entrez Gene ID: | 110253 |
Gene Name: | TRIO and F-actin binding protein |
Gene Aliases: |
AI428493, EST478828, Tara, mKIAA1662 |
Location: |
Chr.15:78947724-79005869 on Build GRCm38 |
Assay Gene Location: | Within Exon 26 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Triobp | NM_001024716.1 | 14 | 2441 | NP_001019887.1 |
| NM_001039156.1 | 25 | 7208 | NP_001034245.1 | |
| NM_138579.4 | 24 | 7070 | NP_613045.3 | |
| AK020187.1 | 4 | 873 | ||
| AK053194.1 | 14 | 2401 | BAC35308.1 | |
| AK085512.1 | 14 | 2412 | ||
| AK129417.1 | 13 | 3866 | BAC98227.2 | |
| AK170390.1 | 14 | 2387 | BAE41764.1 | |
| BC003984.1 | 12 | 2237 | AAH03984.1 | |
| BC042760.1 | 14 | 2380 | AAH42760.1 | |
| BC053329.1 | 14 | 2425 | AAH53329.1 | |
| BP768417.1 | 1 | 159 | ||
| BX528408.1 | 1 | 204 | ||
| BY761337.1 | 2 | 654 | ||
| CX223548.1 | 1 | 105 | ||
| DQ278602.1 | 23 | 6692 | ABB77203.1 |
More Information
Set Membership: |
|

Genomic Map