Assay Details
Target Gene Details
Entrez Gene ID: | 12013 |
Gene Name: | BTB and CNC homology 1 |
Gene Aliases: |
6230421P05Rik, AI323795 |
Location: |
Chr.16:87698954-87733346 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Bach1 | NM_007520.2 | 3 | 840 | NP_031546.1 |
| AK030871.1 | 3 | 790 | ||
| AK140765.1 | 3 | 841 | BAE24472.1 | |
| AK140903.1 | 3 | 841 | BAE24513.1 | |
| AK141405.1 | 3 | 775 | BAE24674.1 | |
| BC057894.1 | 3 | 761 | AAH57894.1 | |
| D86603.1 | 3 | 850 | BAA13137.1 |

Genomic Map