Assay Details
Target Gene Details
Entrez Gene ID: | 319433 |
Gene Name: | serpin peptidase inhibitor, clade E (nexin, plasminogen activator inhibitor type 1), member 3 |
Gene Aliases: |
E130113E03Rik |
Location: |
Chr.14:62663667-62692243 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Serpine3 | NM_001199945.1 | 2 | 142 | NP_001186874.1 |
| AK053602.1 | 2 | 144 |
More Information
Set Membership: |
|

Genomic Map