Assay Details
Target Gene Details
Entrez Gene ID: | 11911 |
Gene Name: | activating transcription factor 4 |
Gene Aliases: |
Atf-4, C/ATF, CREB2, TAXREB67 |
Location: |
Chr.15:80255184-80257545 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Atf4 | NM_001287180.1 | NP_001274109.1 | ||
| NM_009716.3 | 1 | 169 | NP_033846.2 | |
| AA003141.1 | ||||
| AK003001.1 | 1 | 169 | BAC25014.1 | |
| BC085169.1 | 1 | 60 | AAH85169.1 |
More Information
Set Membership: |
|

Genomic Map