Assay Details
Target Gene Details
Entrez Gene ID: | 66335 |
Gene Name: | ATPase, H+ transporting, lysosomal V1 subunit C1 |
Gene Aliases: |
1700025B18Rik, U13839 |
Location: |
Chr.15:38661904-38692444 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Atp6v1c1 | NM_025494.3 | 13 | 1950 | NP_079770.2 |
| AK129007.1 | 10 | 4782 | ||
| AK151237.1 | 13 | 1911 | BAE30229.1 | |
| AK168914.1 | 13 | 1901 | BAE40727.1 | |
| BQ746792.1 | 1 | 392 |
More Information
Set Membership: |
|

Genomic Map