Assay Details
Target Gene Details
Entrez Gene ID: | 110253 |
Gene Name: | TRIO and F-actin binding protein |
Gene Aliases: |
AI428493, EST478828, Tara, mKIAA1662 |
Location: |
Chr.15:78947724-79005869 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Triobp | NM_001039156.1 | 10 | 4360 | NP_001034245.1 |
| NM_138579.4 | 10 | 4360 | NP_613045.3 | |
| AK155197.1 | 3 | 2063 | BAE33111.1 | |
| DQ228000.1 | 10 | 4360 | ABB59556.1 | |
| DQ228001.2 | 10 | 4360 | ABB59557.2 | |
| DQ278602.1 | 9 | 3979 | ABB77203.1 |
More Information
Set Membership: |
|

Genomic Map