Assay Details
Target Gene Details
Entrez Gene ID: | 83925 |
Gene Name: | trichorhinophalangeal syndrome I (human) |
Gene Aliases: |
AI115454, AI447310, D15Ertd586e |
Location: |
Chr.15:50654757-50890747 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Trps1 | NM_032000.2 | 1 | 70 | NP_114389.2 |
| AF346836.1 | 1 | 70 | AAK39508.1 | |
| BC099962.1 | AAH99962.1 |
More Information
Set Membership: |
|

Genomic Map