Assay Details
Target Gene Details
Entrez Gene ID: | 224045 |
Gene Name: | eukaryotic translation initiation factor 2B, subunit 5 epsilon |
Gene Aliases: |
C81315 |
Location: |
Chr.16:20498808-20509325 on Build GRCm38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Eif2b5 | NM_172265.2 | 16 | 2246 | NP_758469.1 |
| AK145801.1 | 16 | 2248 | ||
| AK160985.1 | 16 | 2175 | BAE36132.1 | |
| AK181816.1 | 4 | 317 | ||
| BC038620.1 | 16 | 2246 | AAH38620.1 | |
| BC085255.1 | 16 | 2157 | AAH85255.1 |
More Information
Set Membership: |
|

Genomic Map