Assay Details
Target Gene Details
Entrez Gene ID: | 110920 |
Gene Name: | heat shock protein 70 family, member 13 |
Gene Aliases: |
1600002I10Rik, AV006182, B230217N24Rik, Stch |
Location: |
Chr.16:75755190-75767277 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hspa13 | NM_030201.3 | 5 | 1358 | NP_084477.1 |
| NR_027492.1 | 6 | 1454 | ||
| AK005396.1 | 1 | 195 | ||
| AK030553.1 | 6 | 1427 | ||
| AK034643.1 | 5 | 1338 | BAC28781.1 | |
| AK167839.1 | 5 | 1301 | BAE39860.1 | |
| BC085181.1 | 5 | 1318 | AAH85181.1 |

Genomic Map