Assay Details
Target Gene Details
Entrez Gene ID: | 116731 |
Gene Name: | protocadherin alpha 1 |
Gene Aliases: |
CNR |
Location: |
Chr.18:36930285-37187657 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pcdha1 | NM_054072.1 | NP_473413.1 | ||
| AY013756.1 | AAK26045.1 | |||
| BC082326.1 | AAH82326.1 |
Target Gene Details
Entrez Gene ID: | 353234 |
Gene Name: | protocadherin alpha 2 |
Gene Aliases: |
- |
Location: |
Chr.18:36939205-37187661 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pcdha2 | NM_198117.2 | 1 | 826 | NP_932785.1 |
| AY013760.1 | 1 | 713 | AAK26049.1 | |
| BC030897.1 | 1 | 826 | ||
| BC141390.1 | 1 | 807 |
More Information
Set Membership: |
|

Genomic Map