Assay Details
Target Gene Details
Entrez Gene ID: | 224833 |
Gene Name: | expressed sequence AI661453 |
Gene Aliases: |
- |
Location: |
Chr.17:47436639-47470638 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AI661453 | NM_145489.2 | 5 | 4579 | NP_663464.2 |
| BC029008.1 | 2 | 1388 | AAH29008.1 |

Genomic Map