Assay Details
Target Gene Details
Entrez Gene ID: | 225192 |
Gene Name: | histamine receptor H4 |
Gene Aliases: |
AXOR35, BG26, GPCR105, GPRv53, H4, H4R, HH4R |
Location: |
Chr.18:13006982-13023701 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hrh4 | NM_153087.2 | 1 | 159 | NP_694727.1 |
| AF358859.1 | 1 | 159 | AAK97380.1 | |
| BC111862.1 | 1 | 158 | AAI11863.1 | |
| EU660534.1 | 1 | 112 | ||
| EU660535.1 | 1 | 132 |
More Information
Set Membership: |
|

Genomic Map