Assay Details
Target Gene Details
Entrez Gene ID: | 66053 |
Gene Name: | peptidylprolyl isomerase (cyclophilin)-like 2 |
Gene Aliases: |
0610009L05Rik, 1700016N17Rik, 4921520K19Rik, 4930511F14Rik, AA589416, C130078A06Rik, CYC4 |
Location: |
Chr.16:17086555-17111263 on Build GRCm38 |
Assay Gene Location: | Within Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ppil2 | NM_001252444.1 | 21 | 1958 | NP_001239373.1 |
| NM_144954.3 | 21 | 1860 | NP_659203.1 | |
| AI837084.1 | ||||
| AK012472.1 | 21 | 1939 | ||
| BC028899.1 | 21 | 1821 | AAH28899.1 | |
| M94310.1 | 1 | 155 |
More Information
Set Membership: |
|

Genomic Map