Assay Details
Target Gene Details
Entrez Gene ID: | 11811 |
Gene Name: | apolipoprotein B mRNA editing enzyme, catalytic polypeptide 2 |
Gene Aliases: |
Arp1 |
Location: |
Chr.17:48419231-48432728 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Apobec2 | NM_009694.3 | 2 | 443 | NP_033824.1 |
| AF161699.1 | 2 | 444 | AAD45361.1 | |
| AK009047.1 | 2 | 341 | ||
| BC027530.1 | 2 | 353 | AAH27530.1 |
More Information
Set Membership: |
|

Genomic Map