Assay Details
Target Gene Details
Entrez Gene ID: | 319625 |
Gene Name: | galactose mutarotase |
Gene Aliases: |
A530057M15Rik, AU015645, AU020959 |
Location: |
Chr.17:80127458-80185032 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Galm | NM_176963.4 | 1 | 133 | NP_795937.1 |
| AK040998.1 | 1 | 134 | BAC30776.1 | |
| BC028818.1 | AAH28818.1 |
More Information
Set Membership: |
|

Genomic Map