Assay Details
Target Gene Details
Entrez Gene ID: | 70772 |
Gene Name: | gametogenetin binding protein 1 |
Gene Aliases: |
0610031G08Rik, mdf |
Location: |
Chr.17:27018035-27036378 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ggnbp1 | NM_001251881.1 | 7 | 1004 | NP_001238810.1 |
| NM_027544.2 | 6 | 1433 | NP_081820.2 | |
| AB250379.1 | 6 | 1316 | ||
| AK077129.1 | 6 | 1316 | BAC36632.1 | |
| AY141192.1 | 6 | 1433 | AAN10253.1 | |
| BB018101.1 | 1 | 122 | ||
| BC049764.1 | 6 | 1294 | AAH49764.1 | |
| CO041745.1 | 1 | 109 |
More Information
Set Membership: |
|

Genomic Map