Assay Details
Target Gene Details
Entrez Gene ID: | 104103 |
Gene Name: | antisense Igf2r RNA |
Gene Aliases: |
2810051F02Rik, 2810434M15Rik, AI256653, AI597500, AW049873, Air, B930018I07Rik, D17Ertd663e, IGF2RAS |
Location: |
Chr.17:12741311-12859884 on Build GRCm38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Airn | NR_027773.1 | |||
| AK013242.1 | ||||
| BY718621.1 | ||||
| DQ275620.1 |
Target Gene Details
Entrez Gene ID: | 17171 |
Gene Name: | MAS1 oncogene |
Gene Aliases: |
Mas-1, Mgra |
Location: |
Chr.17:12838841-12868145 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mas1 | NM_008552.4 | 9 | 1660 | NP_032578.2 |
| AK015036.1 | 9 | 1651 | BAB29684.1 | |
| AK030261.1 | 9 | 1650 | BAC26865.1 | |
| AK030265.1 | 9 | 1661 | BAC26868.1 | |
| BC120790.1 | 4 | 982 | ||
| BC125537.1 | 5 | 1045 | ||
| BC144900.1 | 4 | 994 |
More Information
Set Membership: |
|

Genomic Map