Assay Details
Target Gene Details
Entrez Gene ID: | 193813 |
Gene Name: | multiple coagulation factor deficiency 2 |
Gene Aliases: |
1810021C21Rik, F5f8d, Lman1ip, Sdnsf |
Location: |
Chr.17:87254443-87265947 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mcfd2 | NM_139295.3 | 4 | 1135 | NP_647456.1 |
| NM_176808.5 | 5 | 1360 | NP_789778.1 | |
| AF475283.1 | 4 | 1103 | AAM28464.1 | |
| AK033139.1 | 4 | 1069 | BAC28169.1 | |
| AK075781.1 | 5 | 1293 | BAC35955.1 | |
| AK077139.1 | 4 | 1086 | BAC36638.1 | |
| AK077631.1 | 4 | 1069 | BAC36914.1 | |
| AK141599.1 | 5 | 1318 | BAE24757.1 | |
| AK151803.1 | 4 | 1124 | BAE30702.1 | |
| AK215044.1 | 1 | 330 | ||
| BC003996.1 | 4 | 1062 | AAH03996.1 | |
| GQ896355.1 | 3 | 1006 | ||
| GQ896356.1 | 3 | 1006 |
More Information
Set Membership: |
|

Genomic Map