Assay Details
Target Gene Details
Entrez Gene ID: | 13506 |
Gene Name: | desmocollin 2 |
Gene Aliases: |
AW228162, Dsc2a, Dsc2b |
Location: |
Chr.18:20030633-20059552 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dsc2 | NM_013505.3 | 10 | 1561 | NP_038533.1 |
| AK028604.1 | 11 | 1601 | ||
| AK028636.1 | 10 | 1562 | BAC26041.1 | |
| AK030333.1 | 10 | 1562 | BAC26907.1 | |
| AK048547.1 | 10 | 1562 | BAC33371.1 | |
| AK172168.1 | 10 | 1396 | BAE42862.1 | |
| BC004663.1 | 4 | 448 | AAH04663.1 | |
| BC057867.1 | 10 | 1341 | AAH57867.1 | |
| L33779.1 | 10 | 1360 | AAA79176.1 | |
| X73885.1 | 3 | 279 | CAA52089.1 |
More Information
Set Membership: |
|

Genomic Map