Assay Details
Target Gene Details
Entrez Gene ID: | 13511 |
Gene Name: | desmoglein 2 |
Gene Aliases: |
AA408168, D18Ertd293e |
Location: |
Chr.18:20558116-20604526 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dsg2 | NM_007883.2 | 14 | 2404 | NP_031909.2 |
| AB072269.1 | 14 | 2432 | BAB86843.1 | |
| AK029320.1 | 3 | 488 | ||
| AK076345.1 | 2 | 248 | BAC36305.1 | |
| BC034056.1 | 6 | 1194 | AAH34056.1 | |
| BC131653.1 | 14 | 2380 | ||
| BC131654.1 | 14 | 2380 |
More Information
Set Membership: |
|

Genomic Map