Assay Details
Target Gene Details
Entrez Gene ID: | 22061 |
Gene Name: | transformation related protein 63 |
Gene Aliases: |
AI462811, Ket, P51/P63, P63, P73l, Tp63, Trp53rp1 |
Location: |
Chr.16:25683763-25892103 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Trp63 | NM_001127259.1 | 1 | 185 | NP_001120731.1 |
| NM_001127260.1 | 1 | 185 | NP_001120732.1 | |
| NM_001127261.1 | 1 | 185 | NP_001120733.1 | |
| AK138305.1 | 1 | 189 |
More Information
Set Membership: |
|

Genomic Map