Assay Details
Target Gene Details
Entrez Gene ID: | 17287 |
Gene Name: | meprin 1 alpha |
Gene Aliases: |
AI098089, AW107200, Mep-1, Mep-1a, Mep1 |
Location: |
Chr.17:43474324-43504144 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mep1a | NM_008585.2 | 11 | 1283 | NP_032611.2 |
| AK093914.1 | 11 | 1242 | ||
| AK143906.1 | 11 | 1241 | ||
| AK164734.1 | BAE37893.1 | |||
| BC015258.1 | 11 | 1283 | AAH15258.1 | |
| M74897.1 | 11 | 1282 | AAA75354.1 |
More Information
Set Membership: |
|

Genomic Map