Assay Details
Target Gene Details
Entrez Gene ID: | 20016 |
Gene Name: | polymerase (RNA) I polypeptide C |
Gene Aliases: |
40kDa, AA409007, AA959927, AL024089, Polr1e, RPA40, Rpo1-1 |
Location: |
Chr.17:46243920-46248045 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Polr1c | NM_009085.2 | 9 | 1054 | NP_033111.2 |
| AK132114.1 | 9 | 996 | BAE20986.1 | |
| AK146240.1 | 9 | 1004 | BAE27005.1 | |
| BC130018.1 | 9 | 1055 | ||
| CJ207889.1 | 2 | 123 |
More Information
Set Membership: |
|

Genomic Map