Assay Details
Target Gene Details
Entrez Gene ID: | 108077 |
Gene Name: | superkiller viralicidic activity 2-like (S. cerevisiae) |
Gene Aliases: |
4930534J06Rik, AW214248, Ddx13, SKI, Ski2w |
Location: |
Chr.17:34839226-34850204 on Build GRCm38 |
Assay Gene Location: | Within Exon 28 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Skiv2l | NM_021337.2 | 28 | 3833 | NP_067312.2 |
| AK029763.1 | 28 | 3822 | BAC26604.1 | |
| AK087810.1 | 24 | 4469 | BAC40010.1 | |
| AK139051.1 | 26 | 3992 | ||
| AK159854.1 | 28 | 3834 | BAE35430.1 | |
| AK201636.1 | 1 | 88 | ||
| AK206920.1 | 1 | 88 | ||
| AK207240.1 | 1 | 88 | ||
| AK217483.1 | 1 | 88 | ||
| BC023478.1 | 16 | 2437 | AAH23478.1 | |
| BC065999.1 | 28 | 3805 | AAH65999.1 | |
| CF546171.1 | 2 | 410 |

Genomic Map