Assay Details
Target Gene Details
Entrez Gene ID: | 13664 |
Gene Name: | eukaryotic translation initiation factor 1A |
Gene Aliases: |
C76390, Ef1a, Eftu, Eif4c, eIF-1A, eIF-4C |
Location: |
Chr.18:46597658-46610225 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Eif1a | NM_010120.5 | 3 | 2414 | NP_034250.3 |
| AK019451.1 | 3 | 2415 | BAB31727.1 | |
| AK150673.1 | 3 | 2387 | BAE29755.1 | |
| AK150677.1 | 3 | 2387 | BAE29757.1 |
More Information
Set Membership: |
|

Genomic Map