Assay Details
Target Gene Details
Entrez Gene ID: | 108112 |
Gene Name: | eukaryotic translation initiation factor 4E binding protein 3 |
Gene Aliases: |
4e-bp3 |
Location: |
Chr.18:36664060-36666324 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Eif4ebp3 | NM_201256.4 | 3 | 528 | NP_957708.1 |
| AY226182.1 | 3 | 406 | AAO73448.1 | |
| BC061242.1 | 3 | 528 | AAH61242.1 | |
| BC111107.1 | 3 | 468 | AAI11108.1 | |
| CX565744.1 | 2 | 158 |
More Information
Set Membership: |
|

Genomic Map