Assay Details
Target Gene Details
Entrez Gene ID: | 22061 |
Gene Name: | transformation related protein 63 |
Gene Aliases: |
AI462811, Ket, P51/P63, P63, P73l, Tp63, Trp53rp1 |
Location: |
Chr.16:25683763-25892103 on Build GRCm38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Trp63 | NM_001127259.1 | 14 | 3812 | NP_001120731.1 |
| NM_001127260.1 | 13 | 3718 | NP_001120732.1 | |
| NM_001127262.1 | 11 | 3055 | NP_001120734.1 | |
| NM_001127264.1 | 12 | 3137 | NP_001120736.1 | |
| NM_011641.2 | 12 | 3149 | NP_035771.1 | |
| AB010152.1 | 12 | 3123 | BAA32432.1 | |
| AK137253.1 | 12 | 3138 | BAE23286.1 | |
| AK137743.1 | 1 | 641 | ||
| BC090649.1 | 12 | 3119 | AAH90649.1 |
More Information
Set Membership: |
|

Genomic Map