Assay Details
Target Gene Details
Entrez Gene ID: | 75462 |
Gene Name: | RIKEN cDNA 1700001C19 gene |
Gene Aliases: |
- |
Location: |
Chr.17:47412734-47437375 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 1700001C19Rik | NM_001172091.1 | 2 | 759 | NP_001165562.1 |
| NM_029296.2 | 4 | 1041 | NP_083572.1 | |
| BC028263.1 | 4 | 1004 | AAH28263.1 |
Target Gene Details
Entrez Gene ID: | 14913 |
Gene Name: | guanylate cyclase activator 1a (retina) |
Gene Aliases: |
GC-A, Gcap1, Guca1, mGCAP1 |
Location: |
Chr.17:47394558-47413514 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Guca1a |

Genomic Map