Assay Details
Target Gene Details
Entrez Gene ID: | 210148 |
Gene Name: | solute carrier family 30 (zinc transporter), member 6 |
Gene Aliases: |
9530029F08Rik, ZnT6 |
Location: |
Chr.17:74395608-74424229 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc30a6 | NM_001252478.1 | 14 | 1566 | NP_001239407.1 |
| NM_144798.6 | 14 | 1551 | NP_659047.2 | |
| AF233346.1 | AAL83717.1 | |||
| AK082907.1 | 14 | 1528 | BAC38680.1 | |
| AK150342.1 | BAE29482.1 | |||
| AK151120.1 | BAE30129.1 | |||
| AK160903.1 | ||||
| BC005753.1 | AAH05753.1 | |||
| BC066162.1 | 14 | 1518 | AAH66162.1 | |
| BC068169.1 | 14 | 1540 | AAH68169.1 |
More Information
Set Membership: |
|

Genomic Map