Assay Details
Target Gene Details
Entrez Gene ID: | 224674 |
Gene Name: | solute carrier family 37 (glycerol-3-phosphate transporter), member 1 |
Gene Aliases: |
G3PP |
Location: |
Chr.17:31294798-31350709 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc37a1 | NM_153062.2 | 1 | 183 | NP_694702.1 |
| BC027294.1 | 1 | 183 | AAH27294.1 |
More Information
Set Membership: |
|

Genomic Map