Assay Details
Target Gene Details
Entrez Gene ID: | 213760 |
Gene Name: | prolyl endopeptidase-like |
Gene Aliases: |
2810457N15Rik, 9530014L06Rik, D030028O16Rik, mKIAA0436 |
Location: |
Chr.17:85062309-85090274 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Prepl | NM_001163622.1 | 15 | 3173 | NP_001157094.1 |
| NM_001163623.1 | 14 | 2637 | NP_001157095.1 | |
| NM_001163624.1 | 14 | 2505 | NP_001157096.1 | |
| NM_145984.3 | 15 | 2737 | NP_666096.3 | |
| AK083070.1 | 15 | 2678 | BAC38751.1 | |
| BC004612.1 | 14 | 2591 | AAH04612.1 |
Target Gene Details
Entrez Gene ID: | 20532 |
Gene Name: | solute carrier family 3, member 1 |
Gene Aliases: |
D2H, NBAT, NTAA, RBAT |
Location: |
Chr.17:85028347-85064241 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc3a1 | NM_009205.2 | 10 | 2198 | NP_033231.2 |
| AI326967.1 | 1 | 110 | ||
| AK052623.1 | 10 | 2169 | BAC35066.1 | |
| AK132275.1 | 10 | 2170 | BAE21075.1 | |
| BC013441.1 | 10 | 2140 | AAH13441.1 | |
| D88533.1 | 10 | 2158 | BAA24427.1 |
More Information
Set Membership: |
|

Genomic Map