Assay Details
Target Gene Details
Entrez Gene ID: | 277250 |
Gene Name: | KDM3B lysine (K)-specific demethylase 3B |
Gene Aliases: |
5830462I21Rik, JHDM2B, Jmjd1b, mKIAA1082 |
Location: |
Chr.18:34776243-34839370 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kdm3b | NM_001081256.1 | 7 | 1199 | NP_001074725.1 |
| AK129281.1 | BAC98091.1 | |||
| AK147120.1 | 7 | 1142 | BAE27692.1 | |
| AK163281.1 | 8 | 1195 | BAE37274.1 | |
| BC151084.1 | 7 | 1160 |
More Information
Set Membership: |
|

Genomic Map