Assay Details
Target Gene Details
Entrez Gene ID: | 93734 |
Gene Name: | Mpv17 transgene, kidney disease mutant-like |
Gene Aliases: |
M-LP |
Location: |
Chr.16:13903161-13949619 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mpv17l | NM_001289561.1 | 4 | 2662 | NP_001276490.1 |
| NM_001289562.1 | 4 | 2394 | NP_001276491.1 | |
| NM_001289563.1 | 4 | 2357 | NP_001276492.1 | |
| NM_001289565.1 | 5 | 2411 | NP_001276494.1 | |
| NM_001289567.1 | 5 | 2687 | NP_001276496.1 | |
| NM_033564.3 | 4 | 2633 | NP_291042.2 | |
| AF305634.1 | 4 | 2603 | AAK32113.2 | |
| AK136449.1 | 4 | 2395 | BAE22983.1 | |
| BC037713.1 | 4 | 2513 | AAH37713.1 |
More Information
Set Membership: |
|

Genomic Map