Assay Details
Target Gene Details
Entrez Gene ID: | 77037 |
Gene Name: | melanocortin 2 receptor accessory protein |
Gene Aliases: |
1110025G12Rik, Falp, ORF61 |
Location: |
Chr.16:90738324-90749776 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mrap | NM_029844.3 | 3 | 897 | NP_084120.1 |
| AK003912.2 | 3 | 898 | BAB23071.1 | |
| AK158113.1 | 3 | 771 | BAE34361.1 | |
| AY079153.1 | 3 | 674 | AAL80043.1 | |
| BC027543.1 | 3 | 751 | AAH27543.1 |

Genomic Map