Assay Details
Target Gene Details
Entrez Gene ID: | 68172 |
Gene Name: | ribosomal protein L39-like |
Gene Aliases: |
4930517K11Rik |
Location: |
Chr.16:10170192-10174911 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rpl39l | NM_026594.2 | 3 | 590 | NP_080870.1 |
| AK019681.1 | 3 | 592 | BAB31834.1 |
More Information
Set Membership: |
|

Genomic Map