Assay Details
Target Gene Details
Entrez Gene ID: | 68463 |
Gene Name: | mitochondrial ribosomal protein L14 |
Gene Aliases: |
1110006I11Rik, AI846816, MRP-L32, Rpml32 |
Location: |
Chr.17:45685694-45698495 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mrpl14 | NM_026732.2 | 3 | 369 | NP_081008.1 |
| AK003500.1 | 3 | 336 | BAB22824.1 | |
| AK050273.1 | 3 | 355 | BAC34158.1 | |
| AK144153.1 | 3 | 376 | BAE25734.1 | |
| BC027021.1 | 3 | 369 | AAH27021.1 |
More Information
Set Membership: |
|

Genomic Map