Assay Details
Target Gene Details
Entrez Gene ID: | 225363 |
Gene Name: | eukaryotic translation termination factor 1 |
Gene Aliases: |
AI463371, D6Ertd109e, ERF, ERF1, SUP45L1, TB3-1 |
Location: |
Chr.18:34902785-34932003 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Etf1 | NM_144866.3 | 11 | 2949 | NP_659115.3 |
| AK139933.1 | 11 | 2913 |
More Information
Set Membership: |
|

Genomic Map