Assay Details
Target Gene Details
Entrez Gene ID: | 107589 |
Gene Name: | myosin, light polypeptide kinase |
Gene Aliases: |
9530072E15Rik, A930019C19Rik, AW489456, KRP, MLCK108, MLCK210, Mlck, nmMlck, smMLCK |
Location: |
Chr.16:34745199-35002436 on Build GRCm38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mylk | NM_139300.3 | 16 | 2873 | NP_647461.3 |
| AK044527.1 | BAC31966.1 | |||
| AK078556.1 | 1 | 312 | ||
| AY237726.1 | 16 | 2902 | AAO85807.1 | |
| AY237727.2 | 1 | 426 | AAO85808.1 | |
| BC045197.1 | 1 | 356 | AAH45197.1 | |
| BC058610.1 | 16 | 2885 | AAH58610.2 | |
| BC145279.1 | 16 | 2860 | ||
| BC150749.1 | 16 | 2860 |

Genomic Map