Assay Details
Target Gene Details
Entrez Gene ID: | 224055 |
Gene Name: | receptor transporter protein 2 |
Gene Aliases: |
Gm605 |
Location: |
Chr.16:23925548-23930847 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rtp2 | NM_001008230.3 | 1 | 93 | NP_001008231.1 |
| AY562226.1 | AAT70671.1 | |||
| BC049158.1 | 1 | 93 | AAH49158.1 |

Genomic Map