Assay Details
Target Gene Details
Entrez Gene ID: | 69542 |
Gene Name: | RIKEN cDNA 2300002M23 gene |
Gene Aliases: |
Stg, emprin |
Location: |
Chr.17:35567485-35568945 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 2300002M23Rik | NM_175148.3 | 2 | 694 | NP_780357.1 |
| AK009013.1 | 2 | 643 | ||
| AK037046.1 | 2 | 697 | BAC29684.1 | |
| BC107265.1 | 1 | 907 | AAI07266.1 | |
| BC107266.1 | 2 | 694 | AAI07267.1 |

Genomic Map