Assay Details
Target Gene Details
Entrez Gene ID: | 17850 |
Gene Name: | methylmalonyl-Coenzyme A mutase |
Gene Aliases: |
D230010K02Rik, Mcm |
Location: |
Chr.17:40934685-40961989 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mut | NM_008650.3 | 13 | 2419 | NP_032676.2 |
| AK012766.1 | 1 | 129 | BAB28454.1 | |
| AK029425.1 | 1 | 183 | BAC26446.1 | |
| AK146309.1 | 13 | 2379 | BAE27064.1 | |
| AK148304.1 | 2 | 183 | BAE28467.1 | |
| BC019175.1 | 13 | 2401 | AAH19175.1 | |
| X51941.1 | 12 | 2330 | CAA36204.1 |
More Information
Set Membership: |
|

Genomic Map