Assay Details
Target Gene Details
Entrez Gene ID: | 70835 |
Gene Name: | protease, serine 22 |
Gene Aliases: |
4733401N09Rik, BSSP-4, SP001LA |
Location: |
Chr.17:23993534-24000735 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Prss22 | NM_133731.2 | 6 | 989 | NP_598492.2 |
| AB010778.1 | 6 | 972 | BAB20262.1 | |
| AK014645.1 | 6 | 989 | ||
| BC055854.1 | 6 | 974 | AAH55854.1 |
More Information
Set Membership: |
|

Genomic Map