Assay Details
Target Gene Details
Entrez Gene ID: | 107035 |
Gene Name: | F-box protein 38 |
Gene Aliases: |
6030410I24Rik, AU044865, AW214031, Moka, SP329 |
Location: |
Chr.18:62504059-62548858 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fbxo38 | NM_134136.3 | 15 | 2586 | NP_598897.2 |
| AK031096.1 | 15 | 2536 | BAC27250.1 | |
| AK031347.1 | 15 | 2517 | BAC27358.1 | |
| AK034761.1 | 10 | 1773 | BAC28826.1 | |
| AK089046.1 | 15 | 2558 | BAC40718.1 | |
| AK199813.1 | ||||
| AY267463.1 | 15 | 2519 | AAP31028.1 | |
| BC056348.1 | 15 | 2538 | AAH56348.1 |

Genomic Map