Assay Details
Target Gene Details
Entrez Gene ID: | 73694 |
Gene Name: | NADH dehydrogenase (ubiquinone) 1 alpha subcomplex assembly factor 7 |
Gene Aliases: |
2410091C18Rik, AL033374, PRO1853 |
Location: |
Chr.17:78937106-78948052 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ndufaf7 | NM_028611.3 | 10 | 1314 | NP_082887.2 |
| AK010752.1 | 10 | 1313 | BAB27158.2 | |
| AK090097.1 | 9 | 1718 | BAC41089.1 | |
| AK177363.1 | 1 | 198 | ||
| BC028524.1 | 3 | 553 | AAH28524.1 | |
| BC108350.1 | 9 | 1706 | AAI08351.1 | |
| BC116213.1 | 3 | 406 | ||
| BC116214.1 | 3 | 406 | ||
| BC128473.1 | 3 | 406 | ||
| BC128474.1 | 3 | 406 |

Genomic Map