Assay Details
Target Gene Details
Entrez Gene ID: | 213760 |
Gene Name: | prolyl endopeptidase-like |
Gene Aliases: |
2810457N15Rik, 9530014L06Rik, D030028O16Rik, mKIAA0436 |
Location: |
Chr.17:85062309-85090274 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Prepl | NM_001163622.1 | NP_001157094.1 | ||
| NM_001163623.1 | NP_001157095.1 | |||
| NM_001163624.1 | NP_001157096.1 | |||
| NM_145984.3 | NP_666096.3 | |||
| AK020557.1 | 1 | 280 | ||
| AK028877.1 | BAC26168.1 | |||
| AK050878.1 | BAC34440.1 | |||
| AK083070.1 | BAC38751.1 | |||
| AK163108.1 | BAE37195.1 | |||
| BC004612.1 | AAH04612.1 | |||
| BY146774.1 | ||||
| BY257815.1 |
More Information
Set Membership: |
|

Genomic Map