Assay Details
Target Gene Details
Entrez Gene ID: | 63955 |
Gene Name: | CDK5 and Abl enzyme substrate 1 |
Gene Aliases: |
Cables, ik3-1 |
Location: |
Chr.18:11839274-11945627 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cables1 | NM_001146287.1 | 1 | 1053 | NP_001139759.1 |
| NM_022021.2 | 1 | 1053 | NP_071304.2 | |
| AF133208.1 | 1 | 1053 | AAG33933.1 | |
| AF328140.1 | 1 | 738 | AAG42916.1 | |
| AK156273.1 | 1 | 1109 | BAE33652.1 | |
| AK158396.1 | 1 | 783 | BAE34486.1 | |
| BC145142.1 | 1 | 1037 | ||
| BC150714.1 | 1 | 1037 | ||
| CX213994.1 | 1 | 114 |
Target Gene Details
Entrez Gene ID: | 100316686 |
Gene Name: | microRNA 1901 |
Gene Aliases: |
Mirn1901, mmu-mir-1901 |
Location: |
Chr.18:11840361-11840438 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mir1901 | NR_035447.1 |

Genomic Map