Assay Details
Target Gene Details
Entrez Gene ID: | 14164 |
Gene Name: | fibroblast growth factor 1 |
Gene Aliases: |
Dffrx, Fam, Fgf-1, Fgfa |
Location: |
Chr.18:38838672-38929470 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fgf1 | NM_010197.3 | 4 | 1043 | NP_034327.1 |
| AK035330.1 | 4 | 1044 | BAC29035.1 | |
| AK142163.1 | 5 | 1120 | ||
| BC037601.1 | 4 | 1062 | AAH37601.1 | |
| U67610.1 | 4 | 1009 | AAC52907.1 |
More Information
Set Membership: |
|

Genomic Map