Assay Details
Target Gene Details
Entrez Gene ID: | 106369 |
Gene Name: | yippee-like 1 (Drosophila) |
Gene Aliases: |
0610009L05Rik, 1700016N17Rik, 1700019O22Rik, 4921520K19Rik, 4930511F14Rik, AV118478, Dgl1, Ppil2, mdgl-1 |
Location: |
Chr.16:17070138-17086736 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ypel1 | NM_001291047.1 | 3 | 393 | NP_001277976.1 |
| NM_001291053.1 | NP_001277982.1 | |||
| NM_023249.6 | 2 | 267 | NP_075738.1 | |
| AF190624.1 | 2 | 183 | AAG17143.1 | |
| AK005955.1 | 2 | 103 | BAB24338.1 | |
| AK006049.1 | 2 | 103 | BAB24383.2 | |
| AK006137.1 | BAC25133.1 | |||
| AK006505.1 | 2 | 78 | BAB24624.1 | |
| AK006858.1 | 2 | 155 | BAB24767.1 | |
| AK011452.1 | 2 | 199 | ||
| AK014931.1 | 2 | 93 | BAB29625.1 | |
| AK015752.1 | 1 | 103 | BAB29957.1 | |
| AK018910.1 | 2 | 78 | BAB31479.1 | |
| AK019819.1 | 2 | 98 | BAB31864.1 | |
| AK076063.1 | 2 | 210 | BAC36157.1 | |
| AK133401.1 | 2 | 181 | BAE21635.1 | |
| AK139453.1 | 2 | 175 | BAE24018.1 | |
| AK163365.1 | 3 | 334 | ||
| BB610926.1 | 3 | 306 | ||
| BB649357.1 | 2 | 267 | ||
| BC049737.1 | AAH49737.1 | |||
| BQ573264.1 |

Genomic Map