Assay Details
Target Gene Details
Entrez Gene ID: | 74042 |
Gene Name: | RIKEN cDNA 4921501E09 gene |
Gene Aliases: |
PHF8 |
Location: |
Chr.17:33064143-33068058 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 4921501E09Rik | NM_001009544.2 | 1 | 1144 | NP_001009544.1 |
| AK014800.1 | 1 | 1093 | ||
| AK077044.1 | 1 | 1111 | ||
| BC150955.1 | 1 | 991 |

Genomic Map